Merck
  • Mahogunin ring finger 1 suppresses misfolded polyglutamine aggregation and cytotoxicity.
Mahogunin ring finger 1 suppresses misfolded polyglutamine aggregation and cytotoxicity.
Biochimica et biophysica acta (2014-04-29)
Deepak Chhangani, Nobuyuki Nukina, Masaru Kurosawa, Ayeman Amanullah, Vibhuti Joshi, Arun Upadhyay, Amit Mishra
ABSTRACT
Polyglutamine diseases are a family of inherited neurodegenerative diseases caused by the expansion of CAG repeats within the coding region of target genes. Still the mechanism(s) by which polyglutamine proteins are ubiquitinated and degraded remains obscure. Here, for the first time, we demonstrate that Mahogunin 21 ring finger 1 E3 ubiquitin protein ligase is depleted in cells that express expanded-polyglutamine proteins. MGRN1 co-immunoprecipitates with expanded-polyglutamine huntingtin and ataxin-3 proteins. Furthermore, we show that MGRN1 is predominantly colocalized and recruits with polyglutamine aggregates in both cellular and transgenic mouse models. Finally, we demonstrate that the partial depletion of MGRN1 increases the rate of aggregate formation and cell death, whereas the overexpression of MGRN1 reduces the frequency of aggregate formation and provides cytoprotection against polyglutamine-induced proteotoxicity. These observations suggest that stimulating the activity of MGRN1 ubiquitin ligase might be a potential therapeutic target to eliminate the cytotoxic threat in polyglutamine diseases.
Sigma-Aldrich
MISSION® siRNA Universal Negative Control #1
Sigma-Aldrich
MISSION® esiRNA, targeting human MGRN1
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Supelco
2-Mercaptoethanol, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mgrn1