Merck
  • Preparation and friction force microscopy measurements of immiscible, opposing polymer brushes.
Preparation and friction force microscopy measurements of immiscible, opposing polymer brushes.
Journal of visualized experiments : JoVE (2015-01-16)
Sissi de Beer, Edit Kutnyanszky, Martin H Müser, G Julius Vancso
ABSTRACT
Solvated polymer brushes are well known to lubricate high-pressure contacts, because they can sustain a positive normal load while maintaining low friction at the interface. Nevertheless, these systems can be sensitive to wear due to interdigitation of the opposing brushes. In a recent publication, we have shown via molecular dynamics simulations and atomic force microscopy experiments, that using an immiscible polymer brush system terminating the substrate and the slider surfaces, respectively, can eliminate such interdigitation. As a consequence, wear in the contacts is reduced. Moreover, the friction force is two orders of magnitude lower compared to traditional miscible polymer brush systems. This newly proposed system therefore holds great potential for application in industry. Here, the methodology to construct an immiscible polymer brush system of two different brushes each solvated by their own preferred solvent is presented. The procedure how to graft poly(N-isopropylacrylamide) (PNIPAM) from a flat surface and poly(methyl methacrylate) (PMMA) from an atomic force microscopy (AFM) colloidal probe is described. PNIPAM is solvated in water and PMMA in acetophenone. Via friction force AFM measurements, it is shown that the friction for this system is indeed reduced by two orders of magnitude compared to the miscible system of PMMA on PMMA solvated in acetophenone.
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Sigma-Aldrich
Poly(N-isopropylacrylamide), Mn ~40,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Sigma-Aldrich
Poly(methyl methacrylate)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Sigma-Aldrich
Poly(methyl methacrylate)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 mg
Please contact Customer Service for Availability
CA$536.00