- Efficient ring-opening reaction of epoxides and aziridines promoted by tributylphosphine in water.
Efficient ring-opening reaction of epoxides and aziridines promoted by tributylphosphine in water.
The Journal of organic chemistry (2003-02-01)
Ren-Hua Fan, Xue-Long Hou
PMID12558391
ABSTRACT
Tributylphosphine was found to be an effective promoting reagent for the ring-opening reaction of various epoxides and aziridines with nucleophile to produce corresponding anti-bifunctional products in moderate to excellent yields in water.