Merck
  • Comparative analysis of NADPH-diaphorase positive neurons in the rat, rabbit and pheasant thoracic spinal cord. A histochemical study.
Comparative analysis of NADPH-diaphorase positive neurons in the rat, rabbit and pheasant thoracic spinal cord. A histochemical study.
European journal of histochemistry : EJH (2002-01-05)
D Kluchová, S Rybárová, M Miklosová, K Lovásová, K Schmidtová, F Dorko
ABSTRACT
The distribution of NADPH-diaphorase (NADPH-d) activity was investigated and compared in the rat, rabbit and pheasant thoracic spinal cord. The investigation of all spinal cord regions (laminae) in three experimental species revealed marked differences in the distribution of NADPH-d activity. Cross sectional analysis of the spinal cord of the rat, rabbit and pheasant confirmed differences in the shape of the gray matter in all examined species. More detailed investigation of Rexed's laminas showed similar distribution of NADPH-d activity in the spinal cord of the rat and rabbit, which were different when compared with the spinal cord of the pheasant. Ventral horn of the rat and rabbit showed no labelling whereas in pheasant this area possessed a number of scattered, intensively stained neurons. In the location of autonomic preganglionic neurons, differences were found as well. In the rat there was seen a number of densely packed, clearly dark blue coloured neurons. Similarly, these neurons were present in the rabbit spinal cord but they were less numerous. No staining was found in this region of pheasant. Pericentral area (lamina X) and intermediate zone (laminaVII) revealed the presence of NADPH-d positive neurons in all examined species although they differed in number and shape of their bodies. The dorsal horn showed the presence of NADPH-d staining in all three animals but its distribution was different in medio-lateral direction. It can be suggested that observed differencies in the presence and distribution of NADPH-d activity across the examined species may reflect different fylogenetic development.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, ≥93% (HPLC)
Pricing and availability is not currently available.