Merck
  • Identification of mutation in NPC2 by exome sequencing results in diagnosis of Niemann-Pick disease type C.
Identification of mutation in NPC2 by exome sequencing results in diagnosis of Niemann-Pick disease type C.
Molecular genetics and metabolism (2013-06-25)
Afagh Alavi, Shahriar Nafissi, Hosein Shamshiri, Maryam Malakooti Nejad, Elahe Elahi
ABSTRACT
We report identification of a homozygous mutation in NPC2 in two Iranian siblings with a neurologic dysfunction whose disease had not been diagnosed prior to our genetic analysis. The mutation was identified by exome sequencing. The finding resulted in diagnosis of Niemann-Pick disease type C (NPC) in the siblings, and initiation of treatment with Miglustat. The clinical features of the patients are presented. It has been suggested that NPC is under diagnosed, particularly when presentations are not very severe, as was the situation in the cases studied here. NPC is a fatal autosomal recessive disorder clinically characterized by hepatosplenomegaly and progressive neurological deterioration. At the cellular level, it causes aberrant cholesterol trafficking and accumulation of unesterified cholesterol in lysosomes. Mutations in NPC1 and NPC2 are cause of disease in respectively, 95% and 5% of NPC patients. The p.Pro120Ser causing mutation in NPC2 observed in the Iranian patients was earlier observed in the only other NPC2 patient reported from the Middle East. The study demonstrates that in addition to greatly facilitating gene discovery, exome sequencing has notable potentials for diagnosis, particularly for diagnosis of atypical cases.