Merck
  • NHE3 kinase A regulatory protein E3KARP binds the epithelial brush border Na+/H+ exchanger NHE3 and the cytoskeletal protein ezrin.
NHE3 kinase A regulatory protein E3KARP binds the epithelial brush border Na+/H+ exchanger NHE3 and the cytoskeletal protein ezrin.
The Journal of biological chemistry (1998-09-25)
C H Yun, G Lamprecht, D V Forster, A Sidor
ABSTRACT
Cyclic AMP is a major second messenger that inhibits the brush border Na+/H+ exchanger NHE3. We have previously shown that either of two related regulatory proteins, E3KARP or NHERF, is necessary for the cAMP-dependent inhibition of NHE3. In the present study, we characterized the interaction between NHE3 and E3KARP using in vitro binding assays. We found that NHE3 directly binds to E3KARP and that the entirety of the second PSD-95/Dlg/ZO-1 (PDZ) domain plus the carboxyl-terminal domain of E3KARP are required to bind NHE3. E3KARP binds an internal region within the NHE3 C-terminal cytoplasmic tail, defining a new mode of PDZ domain interaction. Analyses of cellular distribution of NHE3 and E3KARP expressed in PS120 fibroblasts show that NHE3 and E3KARP are co-localized on the plasma membrane, but not in a distinct juxtanuclear compartment in which NHE3 is predominantly expressed. The distributions of NHE3 and E3KARP were not affected by treatment with 8-bromo-cAMP. As shown earlier for the human homolog of NHERF, we also found that the cytoskeletal protein ezrin binds to the carboxyl-terminal domain of E3KARP. These results are consistent with the possibility that E3KARP and NHERF may function as scaffold proteins that bind to both NHE3 and ezrin. Since ezrin is a protein kinase A anchoring protein, we suggest that the scaffolding function of E3KARP binding to both ezrin and NHE3 localizes cAMP-dependent protein kinase in the vicinity of the cytoplasmic domain of NHE3, which is phosphorylated by elevated cAMP.