Merck
  • FLICE induced apoptosis in a cell-free system. Cleavage of caspase zymogens.
FLICE induced apoptosis in a cell-free system. Cleavage of caspase zymogens.
The Journal of biological chemistry (1997-01-31)
M Muzio, G S Salvesen, V M Dixit
ABSTRACT
Engagement of CD95 or tumor necrosis factor 1 receptor (TNFR-1) by ligand or agonist antibodies is capable of activating the cell death program, the effector arm of which is composed of mammalian interleukin-1beta converting enzyme (ICE)-like cysteine proteases (designated caspases) that are related to the Caenorhabditis elegans death gene, CED-3. Caspases, unlike other mammalian cysteine proteases, cleave their substrates following aspartate residues. Furthermore, proteases belonging to this family exist as zymogens that in turn require cleavage at internal aspartate residues to generate the two-subunit active enzyme. As such, family members are capable of activating each other. Remarkably, both CD95 and TNFR-1 death receptors initiate apoptosis by recruiting a novel ICE/CED-3 family member, designated FLICE/MACH, to the receptor signaling complex. Therefore, FLICE/MACH represents the apical triggering protease in the cascade. Consistent with this, recombinant FLICE was found capable of proteolytically activating downstream caspases. Furthermore, CrmA, a pox virus-encoded serpin that inhibits Fas and tumor necrosis factor-induced cell death attenuates the ability of FLICE to activate downstream caspases.