Merck
  • Serotonergic reinforcement of intestinal barrier function is impaired in irritable bowel syndrome.
Serotonergic reinforcement of intestinal barrier function is impaired in irritable bowel syndrome.
Alimentary pharmacology & therapeutics (2014-06-20)
D Keszthelyi, F J Troost, D M Jonkers, H M van Eijk, P J Lindsey, J Dekker, W A Buurman, A A M Masclee
ABSTRACT
Alterations in serotonergic (5-HT) metabolism and/or intestinal integrity have been associated with irritable bowel syndrome (IBS). To assess the effects of the precursor of 5-HT, 5-hydroxytryptophan (5-HTP), on mucosal 5-HT availability and intestinal integrity, and to assess potential differences between healthy controls and IBS patients. Fifteen IBS patients and 15 healthy volunteers participated in this randomised double-blind placebo-controlled study. Intestinal integrity was assessed by dual-sugar test and by determining the mucosal expression of tight junction proteins after ingestion of an oral bolus of 100 mg 5-HTP or placebo. Mucosal serotonergic metabolism was assessed in duodenal biopsy samples. 5-HTP administration significantly increased mucosal levels of 5-HIAA, the main metabolite of 5-HT, in both healthy controls (7.1 ± 1.7 vs. 2.5 ± 0.7 pmol/mg, 5-HTP vs. placebo, P = 0.02) and IBS patients (20.0 ± 4.8 vs. 8.1 ± 1.3 pmol/mg, 5-HTP vs. placebo, P = 0.02), with the latter group showing a significantly larger increase. Lactulose/L-rhamnose ratios were significantly lower after administration of 5-HTP (P < 0.05) in healthy controls and were accompanied by redistribution of zonula occludens-1 (ZO-1), pointing to reinforcement of the barrier. In IBS, expression of the tight junction proteins was significantly lower compared to healthy controls, and 5-HTP resulted in a further decrease in occludin expression. Oral 5-HTP induced alterations in mucosal 5-HT metabolism. In healthy controls, a reinforcement of the intestinal barrier was seen whereas such reaction was absent in IBS patients. This could indicate the presence of a serotonin-mediated mechanism aimed to reinforce intestinal barrier function, which seems to dysfunction in IBS patients.
Sigma-Aldrich
5-Hydroxyindole-3-acetic acid, ≥98% (HPLC), crystalline