Merck
Irinotecan and temozolomide brain distribution: a focus on ABCB1.
Cancer chemotherapy and pharmacology (2014-05-29)
Lauriane Goldwirt, Kevin Beccaria, Alexandre Carpentier, Robert Farinotti, Christine Fernandez
ABSTRACT
Glioblastoma (GBM), the most common primary brain tumor in adults, is usually rapidly fatal with median survival duration of only 15 months and a 3-year survival rate of <7 %. Temozolomide (TMZ) is the only anticancer drug that has improved survival in GBM when administered with concomitant radiotherapy. Irinotecan (CPT-11) has also shown efficacy in recurrent gliomas monotherapy with moderate response. As the efficacy of GBM treatments relies on their brain distribution through the blood-brain barrier (BBB), the aim of the present work was to study, on an in vivo model, the brain distribution of TMZ, CPT-11 and its active metabolite, SN-38. We have focussed on the role of ABCB1, the main efflux transporter at the BBB level, through pharmacokinetics studies in CF1 mdr1a(+/+) and mdr1a(-/-) mice. Our results show that TMZ, CPT-11 and SN-38 are transported by ABCB1 at the BBB level with brain/plasma ratios of 1.1, 2.1 and 2.3, respectively.
Product Description
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Theophylline, European Pharmacopoeia (EP) Reference Standard
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Supelco
Temozolomide, VETRANAL®, analytical standard
Supelco
Temozolomide, Pharmaceutical Secondary Standard; Certified Reference Material