Merck
  • Ubiquitin C-terminal hydrolase L1 deletion ameliorates glomerular injury in mice with ACTN4-associated focal segmental glomerulosclerosis.
Ubiquitin C-terminal hydrolase L1 deletion ameliorates glomerular injury in mice with ACTN4-associated focal segmental glomerulosclerosis.
Biochimica et biophysica acta (2014-03-26)
Naomi C Read, Alex Gutsol, Chet E Holterman, Anthony Carter, Josée Coulombe, Douglas A Gray, Chris R J Kennedy
ABSTRACT
Renal ubiquitin C-terminal hydrolase L1 (UCHL1) is upregulated in a subset of human glomerulopathies, including focal segmental glomerulosclerosis (FSGS), where it may serve to promote ubiquitin pools for degradation of cytotoxic proteins. In the present study, we tested whether UCHL1 is expressed in podocytes of a mouse model of ACTN4-associated FSGS. Podocyte UCHL1 protein was detected in glomeruli of K256E-ACTN4(pod+)/UCHL1+/+ mice. UCHL1+/- mice were intercrossed with K256E-ACTN4(pod+) mice and monitored for features of glomerular disease. 10-week-old K256E-ACTN4(pod+)/UCHL1-/- mice exhibited significantly ameliorated albuminuria, glomerulosclerosis, tubular pathology and blood pressure. Interestingly, while UCHL1 deletion diminished both tubular and glomerular apoptosis, WT1-positive nuclei were unchanged. Finally, UCHL1 levels correlated positively with poly-ubiquitinated proteins but negatively with K256E-α-actinin-4 levels, implying reduced K256E-α-actinin-4 proteolysis in the absence of UCHL1. Our data suggest that UCHL1 upregulation in ACTN4-associated FSGS fuels the proteasome and that UCHL1 deletion may impair proteolysis and thereby preserve K256E/wt-α-actinin-4 heterodimers, maintaining podocyte cytoskeletal integrity and protecting the glomerular filtration barrier.
Sigma-Aldrich
MISSION® esiRNA, targeting human UCHL1
Sigma-Aldrich
α-Actinin from chicken gizzard, ~80% α-actinin basis (SDS-PAGE), ammonium sulfate suspension
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Uchl1