Merck
  • Calbindin-D-28k-like immunoreactive structures in the olfactory bulb and anterior olfactory nucleus of the human adult: distribution and cell typology--partial complementarity with parvalbumin.
Calbindin-D-28k-like immunoreactive structures in the olfactory bulb and anterior olfactory nucleus of the human adult: distribution and cell typology--partial complementarity with parvalbumin.
Neuroscience (1991-01-01)
T G Ohm, H Müller, E Braak
ABSTRACT
Calbindin-D-28k and parvalbumin are calcium-binding proteins. The laminar distribution and morphological features of calbindin-D-28k-like immunoreactive structures were studied in 60-microns-thick sections of the human olfactory bulb. Except for the olfactory nerve layer, immunoreactive neurons were present in all layers of the olfactory bulb. They reached highest densities in the external plexiform layer and internal granule cell layer. Considerable numbers of calbindin-like nerve cells were also found in the olfactory tract and in distal portions of the anterior olfactory nucleus. When comparing the distribution of calbindin-positive structures to that of parvalbumin-positive ones a partially complementary distribution pattern was found. Calbindin-like immunoreactive portions of the anterior olfactory nucleus and olfactory tract were mirrored by immunonegative areas in adjacent sections stained for parvalbumin. Using the combined pigment-Nissl procedure we observed the presence of lipofuscin deposits in nearly 80% of all the calbindin-immunoreactive neurons analysed. Moreover, analysis of their lipofuscin deposits rendered the further differentiation of morphologically similar neuronal subpopulations possible. In contrast, all parvalbumin-like immunoreactive neurons remained free of lipofuscin granules.