Merck
  • Respiratory distress and early neonatal lethality in Hspa4l/Hspa4 double-mutant mice.
Respiratory distress and early neonatal lethality in Hspa4l/Hspa4 double-mutant mice.
American journal of respiratory cell and molecular biology (2013-08-29)
Belal A Mohamed, Amal Z Barakat, Torsten Held, Manar Elkenani, Christian Mühlfeld, Jörg Männer, Ibrahim M Adham
ABSTRACT
Heat shock proteins HSPA4L and HSPA4 are closely related members of the HSP110 family and act as cochaperones. We generated Hspa4l(-/-)Hspa4(-/-) mice to investigate a functional complementarity between HSPA4L and HSPA4 during embryonic development. Hspa4l(-/-)Hspa4(-/-) embryos exhibited marked pulmonary hypoplasia and neonatal death. Compared with lungs of wild-type, Hspa4l(-/-), and Hspa4(-/-) embryos, Hspa4l(-/-)Hspa4(-/-) lungs were characterized by diminished saccular spaces and increased mesenchymal septa. Mesenchymal hypercellularity was determined to be due to an increased cell proliferation index and decreased cell death. A significant increase in expression levels of prosurvival protein B cell leukemia/lymphoma 2 may be the cause for inhibition of apoptotic process in lungs of Hspa4(-/-)Hspa4l(-/-) embryos. Accumulation of glycogen and diminished expression of surfactant protein B, prosurfactant protein C, and aquaporin 5 in saccular epithelium suggested impaired maturation of type II and type I pneumocytes in the Hspa4l(-/-)Hspa4(-/-) lungs. Further experiments showed a significant accumulation of ubiquitinated proteins in the lungs of Hspa4l(-/-)Hspa4(-/-) embryos, indicating an impaired chaperone activity. Our study demonstrates that HSPA4L and HSPA4 collaborate in embryonic lung maturation, which is necessary for adaptation to air breathing at birth.