Merck
Sulindac hepatotoxicity: a case report and review.
Journal of toxicology. Clinical toxicology (1985-01-01)
A G Gallanosa, D A Spyker
ABSTRACT
A 44 year old female, previously on propranolol, phenytoin and phenobarbital, developed hepatotoxicity while on sulindac and acetaminophen containing analgesic. A limited review of hepatotoxicity and drug interactions of sulindac is presented. The possible mechanism of hepatotoxicity and its treatment is suggested.
Product Description
Sulindac, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sulindac, ≥98.0%