Merck
[Therapeutic drug monitoring of oxcarbazepine].
Therapie (2010-03-09)
Régis Bouquié, Eric Dailly, Danièle Bentué-Ferrer
ABSTRACT
Oxcarbazepine is an analogue of carbamazepine, used for the treatment of partial seizure with or without secondary generalization. The two forms R and S of the mono-hydroxylated derivatives (MHD) are responsible for most of the anti-convulsant activity and it is the concentrations of MHD that are relevant in therapeutic drug monitoring (TDM). Analysis of currently literature provides no well-established relationship between plasma concentration of MHD and efficiency or toxicity. Although there is not a validated therapeutic range, the residual concentrations of usually observed therapeutic MHD are situated between 12 and 30 mg/L. In certain pathological or physiological circumstances, the pharmacokinetic variability of the oxcarbazepine can be considerable, but this strong unpredictability does not nevertheless justify the TDM of the MHD. Based on the available evidence, TDM of MHD is not routinely warranted but may be possibly useful in specific situations such as pregnancy or renal insufficiency.
Product Description
USP
Oxcarbazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oxcarbazepine, ≥98% (HPLC), solid
Supelco
Oxcarbazepine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Oxcarbazepine, European Pharmacopoeia (EP) Reference Standard