Merck
  • Ledipasvir and sofosbuvir for 8 or 12 weeks for chronic HCV without cirrhosis.
Ledipasvir and sofosbuvir for 8 or 12 weeks for chronic HCV without cirrhosis.
The New England journal of medicine (2014-04-12)
Kris V Kowdley, Stuart C Gordon, K Rajender Reddy, Lorenzo Rossaro, David E Bernstein, Eric Lawitz, Mitchell L Shiffman, Eugene Schiff, Reem Ghalib, Michael Ryan, Vinod Rustgi, Mario Chojkier, Robert Herring, Adrian M Di Bisceglie, Paul J Pockros, G Mani Subramanian, Di An, Evguenia Svarovskaia, Robert H Hyland, Phillip S Pang, William T Symonds, John G McHutchison, Andrew J Muir, David Pound, Michael W Fried
ABSTRACT
High rates of sustained virologic response were observed among patients with hepatitis C virus (HCV) infection who received 12 weeks of treatment with the nucleotide polymerase inhibitor sofosbuvir combined with the NS5A inhibitor ledipasvir. This study examined 8 weeks of treatment with this regimen. In this phase 3, open-label study, we randomly assigned 647 previously untreated patients with HCV genotype 1 infection without cirrhosis to receive ledipasvir and sofosbuvir (ledipasvir-sofosbuvir) for 8 weeks, ledipasvir-sofosbuvir plus ribavirin for 8 weeks, or ledipasvir-sofosbuvir for 12 weeks. The primary end point was sustained virologic response at 12 weeks after the end of therapy. The rate of sustained virologic response was 94% (95% confidence interval [CI], 90 to 97) with 8 weeks of ledipasvir-sofosbuvir, 93% (95% CI, 89 to 96) with 8 weeks of ledipasvir-sofosbuvir plus ribavirin, and 95% (95% CI, 92 to 98) with 12 weeks of ledipasvir-sofosbuvir. As compared with the rate of sustained virologic response in the group that received 8 weeks of ledipasvir-sofosbuvir, the rate in the 12-week group was 1 percentage point higher (97.5% CI, -4 to 6) and the rate in the group that received 8 weeks of ledipasvir-sofosbuvir with ribavirin was 1 percentage point lower (95% CI, -6 to 4); these results indicated noninferiority of the 8-week ledipasvir-sofosbuvir regimen, on the basis of a noninferiority margin of 12 percentage points. Adverse events were more common in the group that received ribavirin than in the other two groups. No patient who received 8 weeks of only ledipasvir-sofosbuvir discontinued treatment owing to adverse events. Ledipasvir-sofosbuvir for 8 weeks was associated with a high rate of sustained virologic response among previously untreated patients with HCV genotype 1 infection without cirrhosis. No additional benefit was associated with the inclusion of ribavirin in the regimen or with extension of the duration of treatment to 12 weeks. (Funded by Gilead Sciences; ION-3 ClinicalTrials.gov number, NCT01851330.).
Sigma-Aldrich
Ribavirin, antiviral
Ribavirin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Uridine 5′-monophosphate, ≥98%