Merck
  • Molecular cloning and characterization of human Aph2 gene, involved in AP-1 regulation by interaction with JAB1.
Molecular cloning and characterization of human Aph2 gene, involved in AP-1 regulation by interaction with JAB1.
Biochimica et biophysica acta (2006-11-25)
Fengrui Zhang, Yujun Di, Jinjun Li, Yan Shi, Liping Zhang, Chunyan Wang, Xianghuo He, Yongzhong Liu, Dafang Wan, Keke Huo, Jianren Gu
ABSTRACT
A human Aph2 gene (hAph2) was identified and cloned from a human placenta cDNA library. Bioinformatics analysis revealed hAPH2 protein shares 96% identity with mouse APH2 and contains a zf-DHHC domain (148-210aa), which is always involved in protein-protein or protein-DNA interaction. Differential expression patterns of hAph2 mRNA were observed in normal human tissues. Yeast two-hybrid screening found another hAPH2-interacting protein JAB1. The zf-DHHC domain of hAPH2 and the C-terminal of JAB1 were confirmed to be critical for the interaction. Fused with GFP and expressed in COS-7, NIH/3T3 and SMMC-7721 cell lines, hAPH2 showed predominant distribution in the cytoplasm and co-localized with JAB1 around the nucleus. Furthermore, overexpression of hAPH2 could increase apoptosis of COS-7 cells and negatively regulate JAB1-induced activation of AP-1 in a concentration dependent manner. The expression level of c-jun was also down-regulated by overexpression of hAPH2 in COS-7 cells. These data showed some basic characterization and function of hAph2 (hAPH2), dependent or independent with JAB1.