Merck
  • [Immunization with live attenuated A/H5N1 vaccine protects guinea pigs from reinfection].
[Immunization with live attenuated A/H5N1 vaccine protects guinea pigs from reinfection].
Voprosy virusologii (2014-04-30)
M V Sergeeva, V V Kuznetsov, M A Kas'ianenko, A A Shaldzhian, I A Repko, M V Potapchuk, Iu R Romanova, L M Tsybalova
ABSTRACT
Cold-adapted influenza virus A/HK/1/68/162/35(H3N2) was developed as unified donor of attenuation and high reproductive capacity forvaccine strains. The reassortant of this donor with surface antigens of highly pathogenic strain Alchicken/Astana/6/05 (H5N1) was tested in guinea pigs as a live or inactivated preparation. Immunization with both formulations induced equal levels of serum virus specific antibodies, while the level of mucosal antibodies was significantly higher in animals immunized with live virus. The challenge with the homologous virus demonstrated complete virus clearance only in this group, thereby indicating a direct correlation of the protection level with the level of mucosal antibodies.
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS