Merck
  • Effects of resveratrol on obesity-related inflammation markers in adipose tissue of genetically obese rats.
Effects of resveratrol on obesity-related inflammation markers in adipose tissue of genetically obese rats.
Nutrition (Burbank, Los Angeles County, Calif.) (2013-09-10)
Saioa Gómez-Zorita, Alfredo Fernández-Quintela, Arrate Lasa, Elizabeth Hijona, Luis Bujanda, María P Portillo
ABSTRACT
The aim of this study was to examine whether resveratrol might represent a promising therapeutic tool with which to combat adipose tissue chronic inflammation in a model of genetic obesity and to link its anti-inflammatory activity with its effect on body fat reduction. Twenty 6-wk-old male Zucker (fa/fa) rats were randomly distributed into two experimental groups. Resveratrol (RSV) was given orally (15 mg/kg body weight/d in RSV group) by means of an orogastric catheter for 6 wk. Enzyme activities were measured spectrophotometrically or fluorimetrically. Gene and protein expressions were analyzed by reverse transcriptase polymerase chain reaction and Western blot respectively. Cytokine concentrations and the activity of nuclear factor κ-light-chain-enhancer of activated β cells (NF-κB) were measured by using commercial kits. RSV reduced the weight of internal adipose tissues. In epididymal depot glucose-6P-dehydrogenase, acetyl-CoA carboxylase activities, as well as lipoprotein lipase expression and activity were reduced by RSV. The expression of hormone-sensitive lipase was increased, and that of the cluster of differentiation 36 was reduced. Serum concentrations of tumor necrosis factor-α, monocyte chemoattractant protein 1, and C-reactive protein were lower in the RSV-treated group than in the control group. Protein expression of interleukin-6 and the activity of NF-κB, were decreased by RSV. The present results provide evidence that fatty acid uptake and lipolysis are metabolic pathways involved in the response of adipose tissue to RSV. This polyphenol modulates plasma cytokine levels partially by reducing macrophage infiltration in adipose tissue and inhibiting NF-κB activity.
Sigma-Aldrich
Acetyl-CoA Carboxylase 1 human, recombinant, expressed in Sf9 cells
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells