Merck
  • Generation of a miniaturized free-flow electrophoresis chip based on a multi-lamination technique--isoelectric focusing of proteins and a single-stranded DNA fragment.
Generation of a miniaturized free-flow electrophoresis chip based on a multi-lamination technique--isoelectric focusing of proteins and a single-stranded DNA fragment.
Analytical and bioanalytical chemistry (2011-09-14)
Britta Walowski, Wilhelm Hüttner, Hainer Wackerbarth
ABSTRACT
Free-flow electrophoresis techniques have been applied for separations in various areas of chemistry and biochemistry. Here we focus on the generation of a free-flow electrophoresis chip and direct monitoring of the separation of different molecules in the separation bed of the miniaturized chip. We demonstrate a fast and efficient way to generate a low-cost micro-free-flow electrophoresis (μFFE) chip with a filling capacity of 9.5 μL based on a multi-lamination technique. Separating webs realized by two transfer-adhesive tapes avoid the problem of gas bubbles entering the separation area. The chip is characterized by isoelectric focusing markers (IEF markers). The functionality of the chip is demonstrated by free-flow isoelectric focusing (FFIEF) of the proteins BSA (bovine serum albumin) and avidin and a single-stranded DNA (ssDNA) fragment in the pH range 3 to 10. The separation voltage ranges between 167 V cm(-1) and 422 V cm(-1), depending on the application.
Sigma-Aldrich
Atto 488-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 488 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488 maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 488 amine, BioReagent, ≥90% (HPLC), suitable for fluorescence
Sigma-Aldrich
Atto 488 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)