Merck
  • COPD patient satisfaction with ipratropium bromide/albuterol delivered via Respimat: a randomized, controlled study.
COPD patient satisfaction with ipratropium bromide/albuterol delivered via Respimat: a randomized, controlled study.
International journal of chronic obstructive pulmonary disease (2013-05-10)
Gary T Ferguson, Mo Ghafouri, Luyan Dai, Leonard J Dunn
ABSTRACT
Ipratropium bromide/albuterol Respimat inhaler (CVT-R) was developed as an environmentally friendly alternative to ipratropium bromide/albuterol metered-dose inhaler (CVT-MDI), which uses a chlorofluorocarbon propellant. The objective of this study was to evaluate patient satisfaction, device usage, and long-term safety of CVT-R compared to CVT-MDI, and to the simultaneous administration of ipratropium bromide hydrofluoroalkane (HFA; I) and albuterol HFA (A) metered-dose inhalers as dual monotherapies (I + A). This is a 48-week, open-label, randomized, active-controlled, parallel-group study (n = 470) comparing CVT-R to CVT-MDI and to I + A. Patients were at least 40 years of age, diagnosed with chronic obstructive pulmonary disease (COPD), and current or exsmokers. Patients were randomized to receive: (1) CVT-R, one inhalation four times daily (QID); or (2) CVT-MDI, two inhalations QID; or (3) I + A two inhalations of each inhaler QID. Patient Satisfaction and Preference Questionnaire (PASAPQ) performance score (primary endpoint) and adverse events. PASAPQ performance score was significantly higher (CVT-R versus CVT-MDI, 9.6; and CVT-R versus I + A, 6.2; both P < 0.001) when using CVT-R compared to CVT-MDI or I + A at all visits starting from week 3, while CVT-MDI and I + A treatment groups were similar. Time to first COPD exacerbation was slightly longer in the CVT-R group compared to the other treatment groups, although it did not reach statistical significance (CVT-R versus CVT-MDI, P = 0.57; CVT-R versus I + A, P = 0.22). Rates of withdrawal and patient refusal to continue treatment were lower in CVT-R compared with CVT-MDI and I + A groups (CVT-R versus CVT-MDI, P = 0.09; CVT-R versus I + A, P = 0.005). The percentage of patients reporting adverse events and serious adverse events was similar across all three treatment groups. CVT-R is an effective, environmentally friendly inhaler that provides patients with a high level of user satisfaction and may positively impact clinical outcomes while having no adverse impacts on patients using the device.
Product Description
Ipratropium bromide impurity B, European Pharmacopoeia (EP) Reference Standard