Merck
  • Hypersensitivity reactions to anticancer agents: data mining of the public version of the FDA adverse event reporting system, AERS.
Hypersensitivity reactions to anticancer agents: data mining of the public version of the FDA adverse event reporting system, AERS.
Journal of experimental & clinical cancer research : CR (2011-10-06)
Kaori Kadoyama, Akiko Kuwahara, Motohiro Yamamori, J B Brown, Toshiyuki Sakaeda, Yasushi Okuno
ABSTRACT
Previously, adverse event reports (AERs) submitted to the US Food and Drug Administration (FDA) database were reviewed to confirm platinum agent-associated hypersensitivity reactions. The present study was performed to confirm whether the database could suggest the hypersensitivity reactions caused by anticancer agents, paclitaxel, docetaxel, procarbazine, asparaginase, teniposide, and etoposide. After a revision of arbitrary drug names and the deletion of duplicated submissions, AERs involving candidate agents were analyzed. The National Cancer Institute Common Terminology Criteria for Adverse Events version 4.0 was applied to evaluate the susceptibility to hypersensitivity reactions, and standardized official pharmacovigilance tools were used for quantitative detection of signals, i.e., drug-associated adverse events, including the proportional reporting ratio, the reporting odds ratio, the information component given by a Bayesian confidence propagation neural network, and the empirical Bayes geometric mean. Based on 1,644,220 AERs from 2004 to 2009, the signals were detected for paclitaxel-associated mild, severe, and lethal hypersensitivity reactions, and docetaxel-associated lethal reactions. However, the total number of adverse events occurring with procarbazine, asparaginase, teniposide, or etoposide was not large enough to detect signals. The FDA's adverse event reporting system, AERS, and the data mining methods used herein are useful for confirming drug-associated adverse events, but the number of co-occurrences is an important factor in signal detection.
Sigma-Aldrich
Teniposide, ≥97% (HPLC)