Merck
  • [Experience with use of immunomodulators in treatment of tonsillitis developed at the background of preimmunization by TEOVac].
[Experience with use of immunomodulators in treatment of tonsillitis developed at the background of preimmunization by TEOVac].
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic] (2011-09-15)
V P Bondarev, S A Mel'nikov, E E Shagarov, N K Chernikova, V I Gutorova, T V Dorokhina, T A Bondareva
ABSTRACT
The use of immunomodulators in the treatment of subjects with postvaccinal reactions to TEOVac was investigated. The most effective schemes were shown to be those with the use of viferon or combination of arbidol and licopide. The terms of the response signs cupping off were much shorter vs. the cases treated with polyoxidonium. The immunomodulating factors did not affect the intensity of the immunity to the vaccine virus.
Sigma-Aldrich
Arbidol hydrochloride, ≥98% (HPLC)