Merck
  • Spectroscopy of the breaking bond: the diradical intermediate of the ring opening in oxazole.
Spectroscopy of the breaking bond: the diradical intermediate of the ring opening in oxazole.
Physical chemistry chemical physics : PCCP (2014-01-22)
Lori M Culberson, Adam A Wallace, Christopher C Blackstone, Dmitry Khuseynov, Andrei Sanov
ABSTRACT
Bond breaking is a challenging problem in both experimental and theoretical chemistry, due to the transient nature and multi-configurational electronic structure of dissociating molecules. We use anion photodetachment to probe the diradical interactions in the ring-opening reaction of oxazole and obtain a self-consistent picture of the breaking bond. Starting from the closed-shell cyclic molecule, the reaction is launched on the anion potential, as an attached electron cleaves a carbon-oxygen bond. In the photodetachment, two neutral potential regions are sampled. One corresponds to a completely dissociated bond, while the other - to the bond fragments separated by approximately 3 Å. At this chemically relevant distance, signatures of lingering through-space interactions between the radical centers are observed.
Sigma-Aldrich
Oxazole, 98%