Merck
  • Synthesis of 2,6-disubstituted dihydropyrans via an efficient BiBr(3)-initiated three component, one-pot cascade.
Synthesis of 2,6-disubstituted dihydropyrans via an efficient BiBr(3)-initiated three component, one-pot cascade.
Tetrahedron (2010-02-18)
Robert J Hinkle, Yajing Lian, Lee C Speight, Heather E Stevenson, Melissa M Sprachman, Lauren A Katkish, M Christa Mattern
ABSTRACT
The rapid synthesis of cis-2,6-disubstituted dihydropyrans is achieved in a three-component, one-pot cascade reaction. BiBr(3)-ediated addition of ketene silyl acetals or silyl enol ethers to beta,gamma-unsaturated cis-4-trimethylsilyl-3-butenal provides a Mukaiyama aldol adduct containing a vinylsilane moiety tethered to a silyl ether. Addition of a second aldehyde initiates a domino sequence involving intermolecular addition followed by an intramolecular silyl-modified Sakurai (ISMS) reaction. Isolated yields of this one-pot reaction vary from 44 to 80% and all compounds were isolated as the cis-diastereomers (10 examples).
Sigma-Aldrich
1-Phenyl-1-trimethylsiloxyethylene, 98%