Merck
  • Metal-free direct oxidative intermolecular diarylation of anilides at ambient temperature assisted by cascade selective formation of C-C and C-N bonds.
Metal-free direct oxidative intermolecular diarylation of anilides at ambient temperature assisted by cascade selective formation of C-C and C-N bonds.
Chemical communications (Cambridge, England) (2012-02-15)
Rajarshi Samanta, Jonas Lategahn, Andrey P Antonchick
ABSTRACT
A new atom-economical process of direct oxidative intermolecular functionalization of aniline derivatives by simple arenes was developed. The products were formed in a highly regioselective manner under metal-free conditions at ambient temperature.
Sigma-Aldrich
1-tert-Butyl-3,5-dimethylbenzene, 98%