Merck
  • Structural analysis of the calcium-binding protein calmodulin from Ascaris suum obliquely striated muscle.
Structural analysis of the calcium-binding protein calmodulin from Ascaris suum obliquely striated muscle.
The Journal of parasitology (1986-04-01)
R A Masaracchia, T C Hassell, M J Donahue
ABSTRACT
Calmodulin was purified from the obliquely striated skeletal muscle of Ascaris suum. The calmodulin had a molecular weight of 16,400 and the amino acid composition indicated it is highly similar to other purified calmodulins, showing insignificant variation in 12 of 17 residues. In the residues that showed variation, a trend towards conservative substitution was observed. Spectrophotometric absorption maxima of 276 nm and 283 nm were observed. A molar absorption coefficient of 7,800 was calculated. Calcium-dependent binding to phenothiazine Affi-Gel confirmed that calcium binding induces conformation changes characteristic of calmodulin. Double reciprocal analysis of phosphodiesterase activation by A. suum calmodulin gave a Kapp of 40 nM.
Sigma-Aldrich
Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder