Merck
  • Reliability and validity of mobile teledermatology in human immunodeficiency virus-positive patients in Botswana: a pilot study.
Reliability and validity of mobile teledermatology in human immunodeficiency virus-positive patients in Botswana: a pilot study.
JAMA dermatology (2014-03-14)
Rahat S Azfar, Robert A Lee, Leslie Castelo-Soccio, Martin S Greenberg, Warren B Bilker, Joel M Gelfand, Carrie L Kovarik
ABSTRACT
Mobile teledermatology may increase access to care. To determine whether mobile teledermatology in human immunodeficiency virus (HIV)-positive patients in Gaborone, Botswana, was reliable and produced valid assessments compared with face-to-face dermatologic consultations. Cross-sectional study conducted in outpatient clinics and public inpatient settings in Botswana for 76 HIV-positive patients 18 years and older with a skin or mucosal condition that had not been evaluated by a dermatologist. We calculated the κ coefficient for diagnosis, diagnostic category, and management for test-retest and interrater reliability. We also determined sensitivity and specificity for each diagnosis. The κ coefficient for test-retest reliability ranged from 0.47 (95% CI, 0.35 to 0.59) to 0.78 (0.67 to 0.88) for the primary diagnosis, 0.29 (0.18 to 0.42) to 0.73 (0.61 to 0.84) for diagnostic category, and 0.17 (-0.01 to 0.36) to 0.54 (0.38 to 0.70) for management. The κ coefficient for interrater reliability ranged from 0.41 (95% CI, 0.31 to 0.52) to 0.51 (0.41 to 0.61) for the primary diagnosis, 0.22 (0.14 to 0.31) to 0.43 (0.34 to 0.53) for diagnostic category, and 0.08 (0.02 to 0.15) to 0.12 (0.01 to 0.23) for management. Sensitivity and specificity for the top 10 diagnoses varied from 0 to 0.88 and 0.84 to 1.00, respectively. Our results suggest that while the use of mobile teledermatology technology in HIV-positive patients in Botswana has significant potential for improving access to care, additional work is needed to improve the reliability and validity of this technology on a larger scale in this population.
Sigma-Aldrich
CKI-7 dihydrochloride, ≥98% (HPLC), solid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
Please contact Customer Service for Availability
€254.00
25 mg
Please contact Customer Service for Availability
€746.00