Merck
  • Dose-dense temozolomide for newly diagnosed glioblastoma: a randomized phase III clinical trial.
Dose-dense temozolomide for newly diagnosed glioblastoma: a randomized phase III clinical trial.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-10-09)
Mark R Gilbert, Meihua Wang, Kenneth D Aldape, Roger Stupp, Monika E Hegi, Kurt A Jaeckle, Terri S Armstrong, Jeffrey S Wefel, Minhee Won, Deborah T Blumenthal, Anita Mahajan, Christopher J Schultz, Sara Erridge, Brigitta Baumert, Kristen I Hopkins, Tzahala Tzuk-Shina, Paul D Brown, Arnab Chakravarti, Walter J Curran, Minesh P Mehta
ABSTRACT
Radiotherapy with concomitant and adjuvant temozolomide is the standard of care for newly diagnosed glioblastoma (GBM). O(6)-methylguanine-DNA methyltransferase (MGMT) methylation status may be an important determinant of treatment response. Dose-dense (DD) temozolomide results in prolonged depletion of MGMT in blood mononuclear cells and possibly in tumor. This trial tested whether DD temozolomide improves overall survival (OS) or progression-free survival (PFS) in patients with newly diagnosed GBM. This phase III trial enrolled patients older than age 18 years with a Karnofsky performance score of ≥ 60 with adequate tissue. Stratification included clinical factors and tumor MGMT methylation status. Patients were randomly assigned to standard temozolomide (arm 1) or DD temozolomide (arm 2) for 6 to 12 cycles. The primary end point was OS. Secondary analyses evaluated the impact of MGMT status. A total of 833 patients were randomly assigned to either arm 1 or arm 2 (1,173 registered). No statistically significant difference was observed between arms for median OS (16.6 v 14.9 months, respectively; hazard ratio [HR], 1.03; P = .63) or median PFS (5.5 v 6.7 months; HR, 0.87; P = .06). Efficacy did not differ by methylation status. MGMT methylation was associated with improved OS (21.2 v 14 months; HR, 1.74; P < .001), PFS (8.7 v 5.7 months; HR, 1.63; P < .001), and response (P = .012). There was increased grade ≥ 3 toxicity in arm 2 (34% v 53%; P < .001), mostly lymphopenia and fatigue. This study did not demonstrate improved efficacy for DD temozolomide for newly diagnosed GBM, regardless of methylation status. However, it did confirm the prognostic significance of MGMT methylation. Feasibility of large-scale accrual, prospective tumor collection, and molecular stratification was demonstrated.
Product Description
Supelco
Temozolomide, VETRANAL®, analytical standard
Sigma-Aldrich
Temozolomide, ≥98% (HPLC)