Merck
  • Inverting sutures for tarsal ectropion (the leicester modified suture technique).
Inverting sutures for tarsal ectropion (the leicester modified suture technique).
Ophthalmic plastic and reconstructive surgery (2013-09-12)
Antonella Berry-Brincat, Joyce Burns, Raghavan Sampath
ABSTRACT
To describe a simple technique to correct tarsal lower eyelid ectropion with or without marked eyelid laxity (The Leicester Modified Suture technique). A retrospective interventional case series of patients undergoing correction of tarsal ectropion with inverting sutures. The study adhered to the principles of the Declaration of Helsinki. Twenty patients (25 eyelids) met the inclusion criteria. The success of the procedure was assessed by improvement of symptoms, eyelid position, and the need for reoperation. The mean postoperative follow up was 3.6 months (range 2-15 months). Eighteen patients (90%) had a successful outcome, while in 2 patients (10%) the outcome was satisfactory, as there was mild residual eversion of the eyelids. None of the patients required reoperation. Tarsal lower eyelid ectropion can be surgically challenging to correct via the transconjunctival plication or subciliary reattachment of retractors. This method is simple to use, resulting in an excellent outcome with no tissue dissection. This technique is easy to learn and implement for oculoplastic surgeons at all levels of experience.
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000