Merck
  • Synthesis and Crystal Structure of a Five-Coordinate Complex of Copper(II) with 4-Nitrobenzenesulfonate and 2, 2'-Bipyridine.
Synthesis and Crystal Structure of a Five-Coordinate Complex of Copper(II) with 4-Nitrobenzenesulfonate and 2, 2'-Bipyridine.
Acta chimica Slovenica (2012-06-01)
Mahboubeh A Sharif, Masoumeh Tabatabaee, Vahideh Beik, Hamid Reza Khavasi
ABSTRACT
[Cu(bipy)2Cl](nbs) (1) (bipy = 2,2'-bipyridine, nbs = 4-nitrobenzenesulfonate) was obtained from the reaction of 4-nitrobenzenesulfonyl chloride and 2-amine-4-methylopyridine with CuCl2 in the presence of 2,2'-bipyridine and characterized by elemental analysis, IR spectra and X-ray single-crystal diffraction. The asymmetric unit of (1) contains the cationic complex [Cu(bipy)2Cl]+ and, in the outer coordination sphere, an (nbs)- counter ion.
Sigma-Aldrich
4-Nitrobenzenesulfonyl chloride, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Nitrobenzenesulfonyl chloride, 97%
Pricing and availability is not currently available.