Merck
Alkyl vs. alkoxy chains on ionic liquid cations.
Chemical communications (Cambridge, England) (2006-10-19)
Wesley A Henderson, Victor G Young, Douglas M Fox, Hugh C De Long, Paul C Trulove
ABSTRACT
The crystal structures and thermal behavior of the 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexa-fluorophosphate salts are compared with the analogous 1-butyl-2,3-dimethylimidazolium salts to examine the influence of the ether oxygen on salt thermal properties for a typical constituent cation used in the preparation of ionic liquids.
Sigma-Aldrich
1,2-Dimethylimidazole, 97%