Merck
  • Synthesis and pharmacological evaluation of novel arylpiperazine derivatives as nonsteroidal androgen receptor antagonists.
Synthesis and pharmacological evaluation of novel arylpiperazine derivatives as nonsteroidal androgen receptor antagonists.
Chemical & pharmaceutical bulletin (2004-11-02)
Isao Kinoyama, Nobuaki Taniguchi, Toru Yoden, Hiroshi Koutoku, Takashi Furutani, Masafumi Kudoh, Minoru Okada
ABSTRACT
The search for novel antiandrogens by high-throughput screening (HTS) of the Yamanouchi chemical library led to the discovery of the lead compound (5), which possesses an arylmorpholine moiety. Through the optimization of the lead compound (5), we have found a series of novel arylpiperazine derivatives. Among them, 4-[4-cyano-(3-trifluoromethyl)phenyl]-N-(4-fluorophenyl)piperazine-1-carboxamide (22; YM-92088) exhibited a potent AR antagonistic activity with an IC(50) value of 0.47 microM in the reporter assay, which is more potent than bicalutamide (4) which has an IC(50) value of 0.89 microM.
Product Description
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm