Merck
  • Liquid chromatographic determination of ethambutol in serum samples based on intramolecular excimer-forming fluorescence derivatization.
Liquid chromatographic determination of ethambutol in serum samples based on intramolecular excimer-forming fluorescence derivatization.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2004-04-08)
Yukitaka Nakano, Hitoshi Nohta, Hideyuki Yoshida, Kenichiro Todoroki, Tetsuya Saita, Hiroshi Fujito, Masato Mori, Masatoshi Yamaguchi
ABSTRACT
A selective liquid chromatographic method has been developed for the assay of ethambutol in serum samples. The assay involves intramolecular excimer-forming derivatization with 4-(1-pyrene)butanoyl chloride (PBC) and isocratic reversed-phase chromatography with fluorescence detection. After acetonitrile-deproteinization of the serum sample, the derivatization reaction of ethambutol with PBC was completed within 30 min at 50 degrees C. N,N'-Diethylethylenediamine was used as an internal standard. The detection limit of ethambutol was 23 ng/ml serum, corresponding to 180 fmol on column at a signal-to-noise ratio of 3. The present method was selective enough to analyze ethambutol in rabbit serum without any tedious sample clean-up procedure because biogenic monoamines gave no peak in the chromatogram. The method was applicable to drug monitoring in rabbit serum.
Sigma-Aldrich
N,N′-Diethylethylenediamine, 95%