Merck
  • Nucleo-cytoplasmic distribution of beta-catenin is regulated by retention.
Nucleo-cytoplasmic distribution of beta-catenin is regulated by retention.
Journal of cell science (2006-03-24)
Eva Krieghoff, Jürgen Behrens, Bernhard Mayr
ABSTRACT
beta-catenin is the central signalling molecule of the canonical Wnt pathway, where it activates target genes in a complex with LEF/TCF transcription factors in the nucleus. The regulation of beta-catenin activity is thought to occur mainly on the level of protein degradation, but it has been suggested that beta-catenin nuclear localization and hence its transcriptional activity may additionally be regulated via nuclear import by TCF4 and BCL9 and via nuclear export by APC and axin. Using live-cell microscopy and fluorescence recovery after photobleaching (FRAP), we have directly analysed the impact of these factors on the subcellular localization of beta-catenin, its nucleo-cytoplasmic shuttling and its mobility within the nucleus and the cytoplasm. We show that TCF4 and BCL9/Pygopus recruit beta-catenin to the nucleus, and APC, axin and axin2 enrich beta-catenin in the cytoplasm. Importantly, however, none of these factors accelerates the nucleo-cytoplasmic shuttling of beta-catenin, i.e. increases the rate of beta-catenin nuclear import or export. Moreover, the cytoplasmic enrichment of beta-catenin by APC and axin is not abolished by inhibition of CRM-1-dependent nuclear export. TCF4, APC, axin and axin2 move more slowly than beta-catenin in their respective compartment, and concomitantly decrease beta-catenin mobility. Together, these data indicate that beta-catenin interaction partners mainly regulate beta-catenin subcellular localization by retaining it in the compartment in which they are localized, rather than by active transport into or out of the nucleus.
Sigma-Aldrich
Escort IV Transfection Reagent, Lipid reagent for transient and stable transfection of mammalian and insect cells.
Pricing and availability is not currently available.