Merck
  • A simplified method for combined immunohistochemistry and in-situ hybridization in fresh-frozen, cryocut mouse brain sections.
A simplified method for combined immunohistochemistry and in-situ hybridization in fresh-frozen, cryocut mouse brain sections.
Brain research. Brain research protocols (2002-07-13)
Sathyanesan Samuel Newton, Antonia Dow, Rose Terwilliger, Ronald Duman
ABSTRACT
A method is described to perform combined immunohistochemistry and in situ hybridization in mouse brain sections. The protocol is specific to sections mounted on glass slides. In contrast to earlier methods that require either paraffin embedding or perfusion of the brain with paraformaldehyde, this protocol can be carried out on fresh-frozen, cryostat cut post-fixed sections. This simple and concise protocol increases the applicability of the technique as the RNAse-free immunodetection of antigen is useful by itself for immunologically identifying specific cells of interest and then examining gene expression in those cells using techniques such as real-time PCR and microarray analysis. The use of fresh-frozen, cryocut sections enables reliable detection of easily perturbable post-translational modifications such as phosphorylation and improves the quality of results obtained in subsequent in situ hybridization by reducing the background signal and interference from lower cell layers. Inducible transgenic mice that express either a dominant negative mutant form of the cAMP response element binding protein (mCREB) or CREB, in discrete brain regions, were used in this study. The combined immunohistochemistry and in situ hybridization protocol was used to examine colocalization of enkephalin or dynorphin mRNA, both downstream targets of CREB-mediated gene expression, in cells expressing transgenic mCREB or CREB.
Sigma-Aldrich
HistoChoice® Clearing Agent, alternative to toluene and xylene