Merck
New metabolites in isovaleric acidemia.
Clinica chimica acta; international journal of clinical chemistry (1981-03-05)
R J Truscott, D Malegan, E McCairns, D Burke, L Hick, P Sims, B Halpern, K Tanaka, L Sweetman, W L Nyhan, J Hammond, C Bumack, E A Haan, D M Danks
ABSTRACT
Two metabolites, 4-hydroxyisovaleric acid and mesaconic acid, have been identified and quantified in the urine of a patient with isovaleric acidemia. These compounds do not appear to have been reported previously as being components of human metabolism. In addition, large quantities of 3-methylbutyrolactone, the lactone of 4-hydroxyisovaleric acid, were observed in the volatile profile obtained by headspace chromatography. The demonstration of 4-hydroxyisovaleric acid supports the contention that urinary methylsuccinic acid seen in patients with isovaleric acidemia has arisen by omega-oxidation of isovaleric acid. The identification of mesaconic acid may indicate that the methylsuccinic acid formed in these patients is subject to further metabolism.
Sigma-Aldrich
Mesaconic acid, 99%