Merck
  • Synthesis and pharmacological evaluation of 1-alkyl-N-[2-ethyl-2-(4-fluorophenyl)butyl]piperidine-4-carboxamide derivatives as novel antihypertensive agents.
Synthesis and pharmacological evaluation of 1-alkyl-N-[2-ethyl-2-(4-fluorophenyl)butyl]piperidine-4-carboxamide derivatives as novel antihypertensive agents.
Bioorganic & medicinal chemistry (2011-08-31)
Susumu Watanuki, Keisuke Matsuura, Yuichi Tomura, Minoru Okada, Toshio Okazaki, Mitsuaki Ohta, Shin-Ichi Tsukamoto
ABSTRACT
We synthesized and evaluated inhibitory activity against T-type Ca(2+) channels for a series of 1-alkyl-N-[2-ethyl-2-(4-fluorophenyl)butyl]piperidine-4-carboxamide derivatives. Structure-activity relationship studies have revealed that dialkyl substituents at the benzylic position play an important role in increasing inhibitory activity. Oral administration of N-[2-ethyl-2-(4-fluorophenyl)butyl]-1-(2-phenylethyl)piperidine-4-carboxamide (20d) lowered blood pressure in spontaneously hypertensive rats without inducing reflex tachycardia, which is often caused by traditional L-type Ca(2+) channel blockers.
Sigma-Aldrich
4-Fluorophenylacetonitrile, 99%
Pricing and availability is not currently available.