Merck
  • Cycloguanil and its parent compound proguanil demonstrate distinct activities against Plasmodium falciparum malaria parasites transformed with human dihydrofolate reductase.
Cycloguanil and its parent compound proguanil demonstrate distinct activities against Plasmodium falciparum malaria parasites transformed with human dihydrofolate reductase.
Molecular pharmacology (1998-12-18)
D A Fidock, T Nomura, T E Wellems
ABSTRACT
The lack of suitable antimalarial agents to replace chloroquine and pyrimethamine/sulfadoxine threatens efforts to control the spread of drug-resistant strains of the malaria parasite Plasmodium falciparum. Here we describe a transformation system, involving WR99210 selection of parasites transformed with either wild-type or methotrexate-resistant human dihydrofolate reductase (DHFR), that has application for the screening of P. falciparum-specific DHFR inhibitors that are active against drug-resistant parasites. Using this system, we have found that the prophylactic drug cycloguanil has a mode of pharmacological action distinct from the activity of its parent compound proguanil. Complementation assays demonstrate that cycloguanil acts specifically on P. falciparum DHFR and has no other significant target. The target of proguanil itself is separate from DHFR. We propose a strategy of combination chemotherapy incorporating the use of multiple parasite-specific inhibitors that act at the same molecular target and thereby maintain, in combination, their effectiveness against alternative forms of resistance that arise from different sets of point mutations in the target. This approach could be combined with traditional forms of combination chemotherapy in which two or more compounds are used against separate targets.