Merck
Inhibition of tumour growth by lipoxygenase inhibitors.
British journal of cancer (1996-09-01)
H J Hussey, M J Tisdale
ABSTRACT
The potential involvement of lipoxygenase metabolites in the tumour growth stimulatory activity of arachidonic and linoleic acid has been studied using the 5-lipoxygenase inhibitors, BWA4C, BWB70C and Zileuton. In vitro the former two agents were relatively potent inhibitors of growth of murine adenocarcinomas (MACs) with IC50 values < 10 microM, whereas Zileuton was less effective. In vivo studies showed BWA4C to be an effective inhibitor of the growth of both the MAC26 and MAC16 tumours at dose levels between 5 and 25 mg kg-1 (b.d.). The growth rate of the MAC26 tumour was also decreased by BWB70C at 25 mg kg-1, whereas lower doses were either ineffective or stimulated tumour growth. This differential effect of the 5-lipoxygenases inhibitors on tumour growth may arise from effects on the 12- and 15-lipoxygenase pathways. To quantify the effect cells were labelled with [3H]arachidonic acid and the biosynthesis of 5-, 12- and 15-hydroxyeicosatetraenoic acid (HETE) was analysed by high-performance liquid chromatography. All three agents caused a decrease in 5-HETE production, although the effect was less pronounced with Zileuton. In MAC26 cells both BWA4C and BWB70C caused a decrease in 12-HETE formation whereas Zileuton had no effect on the other lipoxygenase pathways. The inhibitory effect of these agents on cell growth may result from an imbalance of metabolism of arachidonic acid between the 5-, 12- and 15-lipoxygenase pathways.
Sigma-Aldrich
BW A4C, ≥98% (HPLC)