Merck
  • Activity of subinhibitory concentrations of dapsone alone and in combination with cell-wall inhibitors against Mycobacterium avium complex organisms.
Activity of subinhibitory concentrations of dapsone alone and in combination with cell-wall inhibitors against Mycobacterium avium complex organisms.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (1993-12-01)
N Rastogi, K S Goh, V Labrousse
ABSTRACT
MICs of dapsone (p-p'-diaminodiphenylsulfone) were determined radiometrically for ten strains each of the Mycobacterium avium complex (MAC) and Mycobacterium tuberculosis. MICs ranged from 50 to 250 micrograms/ml for Mycobacterium tuberculosis and from 2 to 100 micrograms/ml for MAC. However, at a concentration as low as 1.5 micrograms/ml dapsone significantly inhibited growth of MAC bacteria when used in combination with other drugs specifically acting at the mycobacterial cell-wall level. The latter drugs were used in subinhibitory concentrations, and included m-fluorophenylalanine (an inhibitor of mycoside-C biosynthesis), ethambutol (an inhibitor of arabinogalactan biosynthesis), and ethionamide (an inhibitor of mycolic acid biosynthesis). Using a radiometric method (Bactec 460-TB), the activity of dapsone was found to be enhanced for 2/10 strains in the presence of m-fluorophenylalanine, for 3/10 strains in the presence of ethambutol and for 5/10 strains in the presence of ethionamide. A satisfactory correlation between the radiometric data and bacterial viable counts was established.
Sigma-Aldrich
m-Fluoro-DL-phenylalanine