MilliporeSigma
Low skin temperatures produced by new skin refrigerants.
The Journal of dermatologic surgery and oncology (1985-12-01)
R A Strick, R L Moy
ABSTRACT
Temperatures produced by Cryosthesia -30 degrees C, Cryosthesia -60 degrees C, and Frigiderm were measured in minipigs. Cryosthesia -60 degrees C and Cryosthesia -30 degrees C were both found to rapidly lower skin temperatures to levels that have been shown to cause cell injury, necrosis, and loss of melanocytes. Use of these agents requires extreme caution in dermabrasion.