Merck
The fate of dienochlor administered orally and dermally to rats.
Toxicology and applied pharmacology (1986-09-15)
G B Quistad, K M Mulholland, W S Skinner
ABSTRACT
Within four days of receiving a single oral dose (1 mg/kg) of [U-ring-14C]dienochlor [bis(pentachloro-2,4-cyclopentadien-1-yl)] female rats excreted 2 and 88% of the applied 14C in urine and feces, respectively. Metabolites could not be identified and the preponderance of the fecal radioactivity consisted of unextractable 14C-labeled residues. Within 1 day virtually all of the dienochlor had been degraded by rats, with only traces of parent dienochlor in excreta and tissues. After four days only 2% of the applied dose remained in tissues (mainly kidney, liver, and gastrointestinal tract). Pharmacokinetic studies with blood plasma and bile showed dienochlor (and/or its metabolites) to be poorly absorbed. Rats were exposed dermally for 24 hr to [14C]dienochlor formulated as Pentac WP miticide both as an aqueous suspension and as an undiluted wettable powder. Half of the dose adhered to the skin and the other half was found in gauze patches used to protect the treated skin. After a 24-hr exposure over 60% of the radiolabel that adhered to skin was removed by washing with an aqueous soap solution and 86% of this rinsing solution was unmetabolized dienochlor. The dienochlor and its metabolites were transported inefficiently from the application site; only 1% of the applied dose was detected in urine plus feces and less than or equal to 0.2% in tissues. With application rates that simulate field exposure by humans, the actual residue of dienochlor and metabolites in skin (i.e., not removable by washing) is about thirteen times higher following exposure to dienochlor as undiluted wettable powder than as an aqueous suspension.