Merck
Pitfall-to-avoid pharyngeal opacities caused by bismuth subgallate.
Canadian Association of Radiologists journal = Journal l'Association canadienne des radiologistes (1989-12-01)
W N Tannous, E M Azouz, T L Tewfik
ABSTRACT
Bismuth subgallate is used occasionally as a hemostatic agent for packing the adenoidal and tonsillar beds after an adenotonsillectomy. Containing a heavy metal, it is relatively insoluble and poorly absorbed. It produces faint opacities on plain radiographs and these may mimic a foreign body or pharyngeal calcification. We report a 13-year-old girl with linear radiopacities in the nasopharynx and in the palatine tonsillar bed. Careful and complete clinical information pinpointed the exact (iatrogenic) cause of these radiodensities.
Sigma-Aldrich
Bismuth(III) gallate basic hydrate, ≥98.0% (calc. on dry substance, KT)