Merck
  • Separation of the enantiomeric intermediates of some platelet-activating factor analogues on a naphthylalanine-type Pirkle column.
Separation of the enantiomeric intermediates of some platelet-activating factor analogues on a naphthylalanine-type Pirkle column.
Journal of chromatography (1990-05-11)
P L Camacho, E Geiger, G Vigh, R Webster, D H Thompson
ABSTRACT
A chiral high-performance liquid-chromatographic separation method was developed for the analysis of some glycerol-based, ether-linked platelet-activating factor precursors. Using a D-naphthylalanine-type Pirkle-column and tetrahydrofuran-hexane eluents, the retention-structure relationships were determined for a large number of glycerol ether derivatives. The derivatives leading to the highest chiral selectivity factors in the analytical separations were surveyed for their applicability in large-scale isolation of chiral synthetic intermediates.