Merck
  • Fine specificity and cross-reactions of monoclonal antibodies to group B streptococcal capsular polysaccharide type III.
Fine specificity and cross-reactions of monoclonal antibodies to group B streptococcal capsular polysaccharide type III.
Vaccine (2012-05-29)
Seth H Pincus, Emily Moran, Grace Maresh, Harold J Jennings, David G Pritchard, Marianne L Egan, Ola Blixt
ABSTRACT
Group B streptococcus (GBS) is a major cause of neonatal sepsis and meningitis. Despite aggressive campaigns using antenatal prophylactic antibiotic therapy, infections continue. Developing an effective maternal vaccine is a public health priority. Antibody (Ab) to the capsular polysaccharide (CPS) is considered the dominant "protective" immune mediator. Here we study the fine specificity and potential host reactivity of a panel of well-characterized murine monoclonal Abs against the type III CPS by examining the binding of the Abs to intact and neuraminidase-digested GBS, purified CPS, synthetic carbohydrate structures, and cells. The results showed marked differences in the fine specificity among these mAbs to a single carbohydrate structure. Cross-reactions with synthetic GD3 and GT3 carbohydrates, representing structures found on surfaces of neural and developing cells, were demonstrated using carbohydrate array technology. The anti-CPS(III) mAbs did not react with cells expressing GD3 and GT3, nor did mAbs specific for the host carbohydrates cross-react with GBS, raising questions about the physiological relevance of this cross-reaction. But in the process of these investigations, we serendipitously demonstrated cross-reactions of some anti-CPS(III) mAbs with antigens, likely carbohydrates, found on human leukocytes. These studies suggest caution in the development of a maternal vaccine to prevent infection by this important human pathogen.
Sigma-Aldrich
Anti-Disialoganglioside GD3 Antibody, clone MB3.6, clone MB3.6, Chemicon®, from mouse