Merck
  • Equal volumes of undiluted nalbuphine and lidocaine and normal diluted saline prevents nalbuphine-induced injection pain.
Equal volumes of undiluted nalbuphine and lidocaine and normal diluted saline prevents nalbuphine-induced injection pain.
Acta anaesthesiologica Taiwanica : official journal of the Taiwan Society of Anesthesiologists (2012-01-10)
Fu-Yuan Wang, Ya-Chun Shen, Mao-Kai Chen, Siu-Wah Chau, Chia-Ling Ku, Yu-Tung Feng, Kuang-I Cheng
ABSTRACT
To determine if the intravenous co-administration of equal volumes of lidocaine and nalbuphine, with undiluted normal saline, prevents injection pain caused by nalbuphine. Eighty adult patients who were scheduled for minor surgeries under general anesthesia delivered via a laryngeal mask airway (LMA) were enrolled in this prospective, randomized, single-blind clinical trial. In the saline group (control) (n = 40), 1 mL (10 mg) nalbuphine was diluted with 9 mL normal saline. In the lidocaine group (experimental) (n = 40), 1 mL (10 mg) nalbuphine was diluted with 1 mL lidocaine (20 mg). The two respective nalbuphine solutions were injected into the cephalic vein at a rate of 20 mL/minute (0.33 mL/second). Pain scores were categorized into five grades. Pain responses upon intravenous injection of nalbuphine, site of cannulation, size of the catheter, and hemodynamic responses to nalbuphine were also recorded. Overall, the median pain score of patients in the lidocaine group was lower than that of the saline group (p < 0.001). In addition, the incidence of injection pain was lower in the lidocaine group than the saline group (2.5% vs. 30%, p = 0.001). A solution of equal volumes of lidocaine and nalbuphine can decrease intravenous nalbuphine-induced injection pain.
Product Description
Supelco
Nalbuphine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nalbuphine hydrochloride hydrate, ≥98% (HPLC)