MilliporeSigma
  • Fluorescence and DNA-binding properties of neodymium(III) and praseodymium(III) complexes containing 1,10-phenanthroline.
Fluorescence and DNA-binding properties of neodymium(III) and praseodymium(III) complexes containing 1,10-phenanthroline.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-15)
Mozhgan Khorasani-Motlagh, Meissam Noroozifar, Sohaila Mirkazehi-Rigi
ABSTRACT
The binding of neodymium(III) and praseodymium(III) complexes containing 1,10-phenanthroline, [M(phen)2Cl3·OH2] (M=Nd (1), Pr (2)), to DNA has been investigated by absorption, emission, and viscosity measurements. The complexes show absorption decreasing in charge transfer band, fluorescence decrement when bound to DNA. The binding constant Kb has been determined by absorption measurement for both complexes and found to be (6.76±0.12)×10(4) for 1 and (1.83±0.15)×10(4)M(-1), for 2. The fluorescence of [M(phen)2Cl3·OH2] (M=Nd (1), Pr (2)) has been studied in detail. The results of fluorescence titration reveal that DNA has the strong ability to quenching the intrinsic fluorescence of Nd(III) and Pr(III) complexes through the static quenching procedure. The binding site number n, apparent binding constant Kb and the Stern-Volmer constant kSV are determined. Thermodynamic parameters, enthalpy change (ΔH°) and entropy change (ΔS°), are calculated according to relevant fluorescent data and Van't Hoff equation. The experimental data suggest that the complexes bind to DNA by non-intercalative mode. Major groove binding is the preferred mode of interaction for [M(phen)2Cl3·OH2] (M=Nd (1), Pr (2)) with DNA.