Merck
  • The interrelationships of glycemic control measures: HbA1c, glycated albumin, fructosamine, 1,5-anhydroglucitrol, and continuous glucose monitoring.
The interrelationships of glycemic control measures: HbA1c, glycated albumin, fructosamine, 1,5-anhydroglucitrol, and continuous glucose monitoring.
Pediatric diabetes (2011-04-19)
Roy Beck, Michael Steffes, Dongyuan Xing, Katrina Ruedy, Nelly Mauras, Darrell M Wilson, Craig Kollman
ABSTRACT
To describe the interrelationships of glycemic control measures: hemoglobin A1c (HbA1c), glycated albumin, fructosamine, 1,5-anhydroglucitrol (1,5-AG), and continuous glucose monitoring (CGM) in children and adolescents with type 1 diabetes. In total, 26 subjects of age 4-17 had HbA1c measurement followed within 14 d by three laboratory measures of glycemia and the collection of CGM glucose data (N = 21). Glycated albumin and fructosamine levels had a higher correlation with each other than with HbA1c. The correlation of 1,5-AG with HbA1c was lower (absolute r value = 0.25). All four measures had a similar degree of correlation with CGM-measured mean glucose (absolute r value = 0.50-0.56) and with hyperglycemic area under the curve (AUC) at 180 mg/dL (0.50-0.60). Each of the four measures (i.e., HbA1c, glycated albumin, fructosamine, and 1,5-AG) had a similar correlation with mean glucose and hyperglycemic AUC-180. 1,5-AG did not correlate with hyperglycemic AUC-180 better than did HbA1c.
Sigma-Aldrich
1,5-Anhydro-D-sorbitol, crystalline, ≥98.0% (TLC)