Merck
Protein kinase affinity reagents based on a 5-aminoindazole scaffold.
Bioorganic & medicinal chemistry letters (2010-11-17)
Ratika Krishnamurty, Amanda M Brock, Dustin J Maly
ABSTRACT
Affinity reagents that target protein kinases are powerful tools for signal transduction research. Here, we describe a general set of kinase ligands based on a 5-aminoindazole scaffold. This scaffold can readily be derivatized with diverse binding elements and immobilized analogs allow selective enrichment of protein kinases from complex mixtures.
Sigma-Aldrich
5-Aminoindazole, 97%