Merck
  • Sensing of human plasma fibrinogen on polished, chemically etched and carbon treated titanium surfaces by diffractive optical element based sensor.
Sensing of human plasma fibrinogen on polished, chemically etched and carbon treated titanium surfaces by diffractive optical element based sensor.
Optics express (2008-07-09)
Raimo Silvennoinen, Vladimir Vetterl, Stanislav Hason, Heikki Tuononen, Martti Silvennoinen, Kari Myller, Ladislav Cvrcek, Jiri Vanek, Patrik Prachar
ABSTRACT
Adsorption of human plasma fibrinogen (HPF) on 6 differently treated titanium samples (polished, polished and etched, and 4 titanium carbide coatings samples produced by using plasma-enhanced chemical vapour deposition (PECVD) method) is investigated by using diffractive optical element (DOE) sensor. Permittivity (susceptibility) change and fluctuation in optical roughness (R(opt)) of treated titanium surface in the presence of background electrolyte without and with HPF molecules are sensed by using DOE sensor and optical ellipsometry. Correlation between transmitted light and thickness of molecule layer was found. The findings allow to sense temporal organization and severity of adsorption of nano-scale HPF molecules on polished, on polished and etched, and on titanium carbide surface.
Sigma-Aldrich
Titanium carbide, nanopowder, ≤200 nm particle size (SEM)
Sigma-Aldrich
Titanium(IV) carbide, −325 mesh, 98%
Sigma-Aldrich
Titanium(IV) carbide, powder, <4 μm, ≥99% (Ti)